Alfa Aesar™ 3,4-Dihydro-2H-1,5-benzodioxepin, 98%

Manufacturer: Alfa Aesar™ H2737406

Quantity 5g

 View more versions of this product

Catalog No. AAH2737406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7216-18-4
    Molecular Formula C9H10O2
    Formula Weight 150.18
    MDL Number MFCD01333457
    Synonym 1,2-Trimethylenedioxybenzene
    Chemical Name or Material 3,4-Dihydro-2H-1,5-benzodioxepin

    Specifications

    Formula Weight 150.18
    Molecular Formula C9H10O2
    MDL Number MFCD01333457
    CAS 7216-18-4
    Synonym 1,2-Trimethylenedioxybenzene
    RTECS Number DF4725200
    Quantity 5g
    Chemical Name or Material 3,4-Dihydro-2H-1,5-benzodioxepin
    Assay 98%

    Description

    Applications
    It is used as an active pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit