Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,4-Dihydro-2H-pyran-5-carboxaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Boiling Point | 90.0°C (15.0 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Synonym | 5,6-dihydro-4h-pyran-3-carbaldehyde, 2h-pyran-5-carboxaldehyde,3,4-dihydro, 2h-pyran-5-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-pyran-5-carbaldehyd, 3,4-dihydro-2h-pyran-5-carboxaldehyde, 3,4-dihydro-2h-pyrane-5-carbaldehyde |
Formula Weight | 112.13 |
Percent Purity | 97% |
Chemical Name or Material | 3,4-Dihydro-2H-pyran-5-carboxaldehyde |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction