Alfa Aesar™ (3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol, 97%

Manufacturer: Alfa Aesar™ H2850606

Quantity 5g

 View more versions of this product

Catalog No. AAH2850606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 304669-35-0
    Molecular Formula C7H14O2Si
    Formula Weight 158.27
    MDL Number MFCD09265116
    Chemical Name or Material (3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol

    Specifications

    Formula Weight 158.27
    Boiling Point 54°C (0.8mmHg)
    Quantity 5g
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Molecular Formula C7H14O2Si
    MDL Number MFCD09265116
    CAS 304669-35-0
    Flash Point 91°C (196°F)
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Chemical Name or Material (3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol
    Assay 97%

    GHS H Statement
    H227
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit