Alfa Aesar™ 3,4-Dihydroxy-DL-phenylalanine, 98%

Manufacturer: Alfa Aesar™ 4153522

Quantity 100g

 View more versions of this product

Catalog No. AA4153522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 63-84-3
    Formula Weight 197.19
    MDL Number MFCD00063060
    Merck Index 14,3420
    Synonym DL-DOPA; 3-Hydroxy-DL-tyrosine
    Chemical Name or Material 3,4-Dihydroxy-DL-phenylalanine

    Specifications

    Formula Weight 197.19
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Physical Form Crystalline
    MDL Number MFCD00063060
    Merck Index 14,3420
    Melting Point 290°C (dec.)
    Quantity 100g
    RTECS Number AY5250000
    CAS 63-84-3
    Synonym DL-DOPA; 3-Hydroxy-DL-tyrosine
    Health Hazard 1 Danger
    Odor Odorless
    Chemical Name or Material 3,4-Dihydroxy-DL-phenylalanine
    Assay 98%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit