Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7027341
Click to view available options
Quantity:
25 g
100 g
500 g
99-50-3
C7H6O4
154.121
MFCD00002509
YQUVCSBJEUQKSH-UHFFFAOYSA-N
protocatechuic acid, 4-carboxy-1,2-dihydroxybenzene, protocatehuic acid, benzoic acid, 3,4-dihydroxy, protocatechuate, 4,5-dihydroxybenzoic acid, 3,4-dihydroxybenzoicacid, unii-36r5qj8l4b, ccris 6291, chembl37537
72
CHEBI:36062
3,4-dihydroxybenzoic acid
C1=CC(=C(C=C1C(=O)O)O)O
99-50-3
C7H6O4
154.121
MFCD00002509
YQUVCSBJEUQKSH-UHFFFAOYSA-N
protocatechuic acid, 4-carboxy-1,2-dihydroxybenzene, protocatehuic acid, benzoic acid, 3,4-dihydroxy, protocatechuate, 4,5-dihydroxybenzoic acid, 3,4-dihydroxybenzoicacid, unii-36r5qj8l4b, ccris 6291, chembl37537
72
CHEBI:36062
3,4-dihydroxybenzoic acid
C1=CC(=C(C=C1C(=O)O)O)O

3,4-Dihydroxybenzoic acid is a metabolite of polyphenols such as phloretin and quercetin. It is also a phenolic acid antioxidant present in fruits, vegetables and nuts. Further, it is found to be an efficacious chemopreventive agent in several carcinogenesis models. In addition to this, it acts as an anti-inflammatory agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydroxybenzoic acid is a metabolite of polyphenols such as phloretin and quercetin. It is also a phenolic acid antioxidant present in fruits, vegetables and nuts. Further, it is found to be an efficacious chemopreventive agent in several carcinogenesis models. In addition to this, it acts as an anti-inflammatory agent.

Solubility
Soluble in hot alcohol and ether. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point ∼202°C (decomposition)
Quantity 25 g
Beilstein 1448841
Merck Index 14,7894
Solubility Information Soluble in hot alcohol and ether. Slightly soluble in water.
Formula Weight 154.12
Percent Purity 97%
Chemical Name or Material 3,4-Dihydroxybenzoic acid
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-760-0
RTECSNumber UL0560000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit