Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydroxybenzonitrile, 98+%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
17345-61-8
C7H5NO2
135.122
MFCD00016436
NUWHYWYSMAPBHK-UHFFFAOYSA-N
protocatachuonitrile,3,4-dihydroxy benzonitrile,3,4-dihydroxybenzenecarbonitrile,protocatechuonitrile,4-cyanocatechol,protocatechunitrile,4-cyanopyrocatechol,pubchem3120,acmc-209e6q,3, 4-dihydroxybenzonitrile
87065
3,4-dihydroxybenzonitrile
C1=CC(=C(C=C1C#N)O)O
17345-61-8
C7H5NO2
135.122
MFCD00016436
NUWHYWYSMAPBHK-UHFFFAOYSA-N
protocatachuonitrile,3,4-dihydroxy benzonitrile,3,4-dihydroxybenzenecarbonitrile,protocatechuonitrile,4-cyanocatechol,protocatechunitrile,4-cyanopyrocatechol,pubchem3120,acmc-209e6q,3, 4-dihydroxybenzonitrile
87065
3,4-dihydroxybenzonitrile
C1=CC(=C(C=C1C#N)O)O
Packaging Glass bottle
Color White
Melting Point 154°C to 157°C
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula (OH)2C6H3CN
Formula Weight 135.12
Physical Form Powder
Percent Purity ≥98.0%
Chemical Name or Material 3, 4-Dihydroxybenzonitrile
Show More Show Less

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning