Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dihydroxybenzylamine hydrobromide, 98%

Click to view available options
Quantity:
1g
5g
25g
16290-26-9
C7H10BrNO2
220.066
MFCD00012859
BVFZTXFCZAXSHN-UHFFFAOYSA-N
3,4-dihydroxybenzylamine hydrobromide,4-aminomethyl benzene-1,2-diol hydrobromide,dhba hydrobromide,acmc-1afa4,1, 4-aminomethyl-, hydrobromide,4-aminomethyl catechol hydrobromide,3,4 dihydroxybenzylamine hydrobromide,3,4-dihy-droxybenzylamine hydrobromide,3,4-dihydroxy benzylamine hydrobromide,4-aminomethyl benzene-1,2-diol, bromide
13343562
4-(aminomethyl)benzene-1,2-diol;hydrobromide
C1=CC(=C(C=C1CN)O)O.Br
16290-26-9
C7H10BrNO2
220.066
MFCD00012859
BVFZTXFCZAXSHN-UHFFFAOYSA-N
3,4-dihydroxybenzylamine hydrobromide,4-aminomethyl benzene-1,2-diol hydrobromide,dhba hydrobromide,acmc-1afa4,1, 4-aminomethyl-, hydrobromide,4-aminomethyl catechol hydrobromide,3,4 dihydroxybenzylamine hydrobromide,3,4-dihy-droxybenzylamine hydrobromide,3,4-dihydroxy benzylamine hydrobromide,4-aminomethyl benzene-1,2-diol, bromide
13343562
4-(aminomethyl)benzene-1,2-diol;hydrobromide
C1=CC(=C(C=C1CN)O)O.Br
Quantity 25g
Melting Point 196°C to 200°C
Beilstein 4002646
Formula Weight 220.07
Percent Purity 98%
Chemical Name or Material 3,4-Dihydroxybenzylamine hydrobromide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning