Alfa Aesar™ 3,4'-Diisopropylbiphenyl, 94%

Manufacturer: Alfa Aesar™ H2739314

Quantity 25g

 View more versions of this product

Catalog No. AAH2739314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 61434-46-6
    Molecular Formula C18H22
    Formula Weight 238.37
    MDL Number MFCD00043539
    Chemical Name or Material 3,4'-Diisopropylbiphenyl

    Specifications

    Formula Weight 238.37
    MDL Number MFCD00043539
    Molecular Formula C18H22
    Quantity 25g
    CAS 61434-46-6
    Chemical Name or Material 3,4'-Diisopropylbiphenyl
    Assay 94%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit