Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dimethoxy-beta-nitrostyrene, 99%

Click to view available options
Quantity:
5g
25g
100g
4230-93-7
C10H11NO4
209.201
MFCD00024823
SYJMYDMKPSZMSB-AATRIKPKSA-N
3,4-dimethoxy-b-nitrostyrene,1,2-dimethoxy-4-2-nitrovinyl benzene,1,2-dimethoxy-4-e-2-nitroethenyl benzene,3,4-dimethoxy-.beta.-nitrostyrene,3,4-dimethoxy-beta-nitrostyrene,3,4-dimethoxy-omega-nitrostyrene,ethylene, 1-3,4-dimethoxy phenyl-2-nitro,3,4-dimethoxynitrostyrene,3,4-dimethoxy-,a-nitrostyrene,1-3,4-dimethoxyphenyl-2-nitroethene
709688
1,2-dimethoxy-4-[(E)-2-nitroethenyl]benzene
COC1=C(C=C(C=C1)C=C[N+](=O)[O-])OC
4230-93-7
C10H11NO4
209.201
MFCD00024823
SYJMYDMKPSZMSB-AATRIKPKSA-N
3,4-dimethoxy-b-nitrostyrene,1,2-dimethoxy-4-2-nitrovinyl benzene,1,2-dimethoxy-4-e-2-nitroethenyl benzene,3,4-dimethoxy-.beta.-nitrostyrene,3,4-dimethoxy-beta-nitrostyrene,3,4-dimethoxy-omega-nitrostyrene,ethylene, 1-3,4-dimethoxy phenyl-2-nitro,3,4-dimethoxynitrostyrene,3,4-dimethoxy-,a-nitrostyrene,1-3,4-dimethoxyphenyl-2-nitroethene
709688
1,2-dimethoxy-4-[(E)-2-nitroethenyl]benzene
COC1=C(C=C(C=C1)C=C[N+](=O)[O-])OC
Melting Point 139°C to 141°C
Quantity 5g
Formula Weight 209.19
Percent Purity 99%
Chemical Name or Material 3,4-Dimethoxy-beta-nitrostyrene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning