Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxy-N-methylbenzylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
63-64-9
C10H15NO2
181.24
XQODFBIAQVJQHF-UHFFFAOYSA-N
3,4-dimethoxybenzyl methylamine,n-methyl-3,4-dimethoxybenzylamine,1-3,4-dimethoxyphenyl-n-methylmethanamine,3,4-dimethoxyphenyl methyl methyl amine,3,4-dimethoxy-n-methylbenzylamine,3,4-dimethoxy-benzyl-methyl-amine,benzenemethanamine, 3,4-dimethoxy-n-methyl,1-3,4-dimethoxyphenyl-n-methyl-methanamine,3,4-dimethoxyphenyl methyl methylamine,3,4-dimethoxybenzyl-n-methylamine
592161
1-(3,4-dimethoxyphenyl)-N-methylmethanamine
CNCC1=CC(=C(C=C1)OC)OC
63-64-9
C10H15NO2
181.24
XQODFBIAQVJQHF-UHFFFAOYSA-N
3,4-dimethoxybenzyl methylamine,n-methyl-3,4-dimethoxybenzylamine,1-3,4-dimethoxyphenyl-n-methylmethanamine,3,4-dimethoxyphenyl methyl methyl amine,3,4-dimethoxy-n-methylbenzylamine,3,4-dimethoxy-benzyl-methyl-amine,benzenemethanamine, 3,4-dimethoxy-n-methyl,1-3,4-dimethoxyphenyl-n-methyl-methanamine,3,4-dimethoxyphenyl methyl methylamine,3,4-dimethoxybenzyl-n-methylamine
592161
1-(3,4-dimethoxyphenyl)-N-methylmethanamine
CNCC1=CC(=C(C=C1)OC)OC
Flash Point 110°C
Color Brown to Colorless
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 181.24
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3, 4-Dimethoxy-N-methylbenzylamine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger