Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dimethoxybenzenesulfonyl chloride, 98%

Click to view available options
Quantity:
5g
25g
100g
23095-31-0
C8H9ClO4S
236.666
MFCD00051769
RSJSYCZYQNJQPY-UHFFFAOYSA-N
3,4-dimethoxybenzene-1-sulfonyl chloride,3,4-dimethoxybenzenesulfonylchloride,3,4-dimethoxyphenylsulfonyl chloride,3,4-dimethoxy benzenesulfonyl chloride,benzenesulfonyl chloride, 3,4-dimethoxy,3,4-dimethoxyphenyl chlorosulfone,pubchem5127,acmc-1co88,asischem d48920,zerenex zx009571
2734183
3,4-dimethoxybenzenesulfonyl chloride
COC1=C(C=C(C=C1)S(=O)(=O)Cl)OC
23095-31-0
C8H9ClO4S
236.666
MFCD00051769
RSJSYCZYQNJQPY-UHFFFAOYSA-N
3,4-dimethoxybenzene-1-sulfonyl chloride,3,4-dimethoxybenzenesulfonylchloride,3,4-dimethoxyphenylsulfonyl chloride,3,4-dimethoxy benzenesulfonyl chloride,benzenesulfonyl chloride, 3,4-dimethoxy,3,4-dimethoxyphenyl chlorosulfone,pubchem5127,acmc-1co88,asischem d48920,zerenex zx009571
2734183
3,4-dimethoxybenzenesulfonyl chloride
COC1=C(C=C(C=C1)S(=O)(=O)Cl)OC
Quantity 5g
Sensitivity Moisture sensitive
Melting Point 66°C to 70°C
UN Number UN3261
Beilstein 2840170
Formula Weight 236.68
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxybenzenesulfonyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures