Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxybenzhydrazide, 98+%, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
41764-74-3
C9H12N2O3
196.206
MFCD00017061
LJMQIGMMUZLDOC-UHFFFAOYSA-N
3,4-dimethoxybenzhydrazide, veratrohydrazide, 3,4-dimethoxybenzenecarbohydrazide, 3,4-dimethoxybenzoic acid hydrazide, 1,2-dimethoxybenzene-4-carbohydrazide, veratric acid hydrazide, acmc-1aq0x, dimethoxybenzenecarbohydrazide, benzoic acid,3,4-dimethoxy-, hydrazide, benzoic acid, 3,4-dimethoxy-, hydrazide
1530859
3,4-dimethoxybenzohydrazide
COC1=C(C=C(C=C1)C(=O)NN)OC
41764-74-3
C9H12N2O3
196.206
MFCD00017061
LJMQIGMMUZLDOC-UHFFFAOYSA-N
3,4-dimethoxybenzhydrazide, veratrohydrazide, 3,4-dimethoxybenzenecarbohydrazide, 3,4-dimethoxybenzoic acid hydrazide, 1,2-dimethoxybenzene-4-carbohydrazide, veratric acid hydrazide, acmc-1aq0x, dimethoxybenzenecarbohydrazide, benzoic acid,3,4-dimethoxy-, hydrazide, benzoic acid, 3,4-dimethoxy-, hydrazide
1530859
3,4-dimethoxybenzohydrazide
COC1=C(C=C(C=C1)C(=O)NN)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 143°C to 146°C
Beilstein 2651843
Quantity 25g
Formula Weight 196.21
Percent Purity ≥98%
Chemical Name or Material 3,4-Dimethoxybenzhydrazide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 255-541-7

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only