Alfa Aesar™ 3,4-Dimethoxybenzoic acid, 99+%

Manufacturer: Alfa Aesar™ A1016718

Quantity 50g

 View more versions of this product

Catalog No. AAA1016718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 93-07-2
    Molecular Formula C9H10O4
    Formula Weight 182.18
    MDL Number MFCD00002500
    Beilstein 518285
    Merck Index 14,9953
    Synonym Veratric acid
    Chemical Name or Material 3,4-Dimethoxybenzoic acid

    Specifications

    Formula Weight 182.18
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 93-07-2
    Quantity 50g
    Synonym Veratric acid
    TSCA TSCA
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number DG8598750
    Melting Point 179° to 183°C
    MDL Number MFCD00002500
    Beilstein 518285
    Merck Index 14,9953
    Molecular Formula C9H10O4
    Health Hazard 1 Warning
    Chemical Name or Material 3,4-Dimethoxybenzoic acid
    Assay ≥99%

    Description

    Applications
    3,4-Dimethoxybenzoic acid plays an important role in producing antibiotics and various dyes. They are used as intermediates for pharmaceuticals (especially for antipyretic analgesic, antirheumatism) and other organic synthesis. It is used as matrix for ionization of peptides, proteins and carbohydrates.

    Solubility
    Soluble in water (0.5 mg/ml at 15°C), chloroform, methanol, and 95% ethanol (50 mg/ml).

    Notes
    Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit