Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxybenzoic acid, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
93-07-2
C9H10O4
182.18
MFCD00002500
DAUAQNGYDSHRET-UHFFFAOYSA-N
veratric acid,benzoic acid, 3,4-dimethoxy,dimethylprotocatechuic acid,veratrylic acid,3,4-dimethylprotocatechuic acid,veratrumenoic acid,3,4-dimethoxy-benzoic acid,unii-1yy04e7rr4,3,4-dimethoxybenzoes
7121
CHEBI:296881
3,4-dimethoxybenzoic acid
COC1=C(C=C(C=C1)C(=O)O)OC
93-07-2
C9H10O4
182.18
MFCD00002500
DAUAQNGYDSHRET-UHFFFAOYSA-N
veratric acid,benzoic acid, 3,4-dimethoxy,dimethylprotocatechuic acid,veratrylic acid,3,4-dimethylprotocatechuic acid,veratrumenoic acid,3,4-dimethoxy-benzoic acid,unii-1yy04e7rr4,3,4-dimethoxybenzoes
7121
CHEBI:296881
3,4-dimethoxybenzoic acid
COC1=C(C=C(C=C1)C(=O)O)OC
Flash Point >100°C
Color Beige to White or Red
Melting Point 179°C to 182°C
Quantity 100g
Infrared Spectrum Authentic
Packaging Glass bottle
Water 0.5% max. (K.F.)
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula (CH3O)2C6H3CO2H
Beilstein 10, I, 188
Merck Index 15, 10147
Solubility Information Solubility in water: 0.5g/L (14°C). Other solubilities: readily soluble in alcohol,soluble in ether
Formula Weight 182.18
Physical Form Crystalline Powder
Percent Purity 99.0 to 101.0%
Chemical Name or Material 3, 4-Dimethoxybenzoic acid
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.