Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dimethoxybenzoyl chloride, 98%

Click to view available options
Quantity:
5g
25g
3535-37-3
C9H9ClO3
200.618
MFCD00000674
VIOBGCWEHLRBEP-UHFFFAOYSA-N
veratroyl chloride,3,4-dimethoxybenzoylchloride,3,4-dimethoxybenzene-1-carbonyl chloride,benzoyl chloride, 3,4-dimethoxy,3,4-dimethoxy benzoyl chloride,3,4-dimethoxy-benzoyl chloride,3,4-dimethoxy-1-benzenecarbonyl chloride,veratroylchlorid,zlchem 1258,ksc225m7t
77070
3,4-dimethoxybenzoyl chloride
COC1=C(C=C(C=C1)C(=O)Cl)OC
3535-37-3
C9H9ClO3
200.618
MFCD00000674
VIOBGCWEHLRBEP-UHFFFAOYSA-N
veratroyl chloride,3,4-dimethoxybenzoylchloride,3,4-dimethoxybenzene-1-carbonyl chloride,benzoyl chloride, 3,4-dimethoxy,3,4-dimethoxy benzoyl chloride,3,4-dimethoxy-benzoyl chloride,3,4-dimethoxy-1-benzenecarbonyl chloride,veratroylchlorid,zlchem 1258,ksc225m7t
77070
3,4-dimethoxybenzoyl chloride
COC1=C(C=C(C=C1)C(=O)Cl)OC
Boiling Point 95°C to 98°C (1mmHg)
Quantity 25g
Odor Irritating
Melting Point 70°C to 72°C
UN Number UN3261
Beilstein 783596
Formula Weight 200.62
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxybenzoyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.