Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dimethoxybenzylamine, 97%

Click to view available options
Quantity:
5g
100g
5763-61-1
C9H13NO2
167.208
MFCD00008116
DIVNUTGTTIRPQA-UHFFFAOYSA-N
3,4-dimethoxybenzylamine,veratrylamine,3,4-dimethoxyphenyl methanamine,benzenemethanamine, 3,4-dimethoxy,1-3,4-dimethoxyphenyl methanamine,benzylamine, 3,4-dimethoxy,3,4-dimethoxyphenyl methylamine,veratrylamin,pubchem7394,acmc-1altc
79832
(3,4-dimethoxyphenyl)methanamine
COC1=C(C=C(C=C1)CN)OC
5763-61-1
C9H13NO2
167.208
MFCD00008116
DIVNUTGTTIRPQA-UHFFFAOYSA-N
3,4-dimethoxybenzylamine,veratrylamine,3,4-dimethoxyphenyl methanamine,benzenemethanamine, 3,4-dimethoxy,1-3,4-dimethoxyphenyl methanamine,benzylamine, 3,4-dimethoxy,3,4-dimethoxyphenyl methylamine,veratrylamin,pubchem7394,acmc-1altc
79832
(3,4-dimethoxyphenyl)methanamine
COC1=C(C=C(C=C1)CN)OC
Density 1.109
Refractive Index 1.556
Sensitivity Air Sensitive
Quantity 5g
Flash Point >110°C (230°F)
Boiling Point 281°C to 284°C
UN Number UN2735
Beilstein 511575
Formula Weight 167.21
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxybenzylamine
Show More Show Less

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 227-287-7

RTECSNumber : YX7355000

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen