Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dimethoxybenzylamine, 97%

Click to view available options
Quantity:
5g
100g
5763-61-1
C9H13NO2
167.208
MFCD00008116
DIVNUTGTTIRPQA-UHFFFAOYSA-N
3,4-dimethoxybenzylamine,veratrylamine,3,4-dimethoxyphenyl methanamine,benzenemethanamine, 3,4-dimethoxy,1-3,4-dimethoxyphenyl methanamine,benzylamine, 3,4-dimethoxy,3,4-dimethoxyphenyl methylamine,veratrylamin,pubchem7394,acmc-1altc
79832
(3,4-dimethoxyphenyl)methanamine
COC1=C(C=C(C=C1)CN)OC
5763-61-1
C9H13NO2
167.208
MFCD00008116
DIVNUTGTTIRPQA-UHFFFAOYSA-N
3,4-dimethoxybenzylamine,veratrylamine,3,4-dimethoxyphenyl methanamine,benzenemethanamine, 3,4-dimethoxy,1-3,4-dimethoxyphenyl methanamine,benzylamine, 3,4-dimethoxy,3,4-dimethoxyphenyl methylamine,veratrylamin,pubchem7394,acmc-1altc
79832
(3,4-dimethoxyphenyl)methanamine
COC1=C(C=C(C=C1)CN)OC
Flash Point >110°C (230°F)
Boiling Point 281°C to 284°C
Quantity 100g
UN Number UN2735
Beilstein 511575
Formula Weight 167.21
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxybenzylamine

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

RTECSNumber : YX7355000