Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxyphenethyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
7417-21-2
C10H14O3
182.22
MFCD00002894
SRQAJMUHZROVHW-UHFFFAOYSA-N
3,4-dimethoxyphenethyl alcohol,2-3,4-dimethoxyphenyl ethanol,3,4-dimethoxyphenethylalcohol,2-3,4-dimethoxyphenyl ethan-1-ol,3,4-dimethoxyphenethanol,homoveratryl alcohol,benzeneethanol, 3,4-dimethoxy,3,4-dimethoxybenzeneethanol,3,4-dimethoxy-.beta.-phenethyl alcohol,1-2-hydroxyethyl-3,4-dimethoxybenzene
81911
2-(3,4-dimethoxyphenyl)ethanol
COC1=C(C=C(C=C1)CCO)OC
7417-21-2
C10H14O3
182.22
MFCD00002894
SRQAJMUHZROVHW-UHFFFAOYSA-N
3,4-dimethoxyphenethyl alcohol,2-3,4-dimethoxyphenyl ethanol,3,4-dimethoxyphenethylalcohol,2-3,4-dimethoxyphenyl ethan-1-ol,3,4-dimethoxyphenethanol,homoveratryl alcohol,benzeneethanol, 3,4-dimethoxy,3,4-dimethoxybenzeneethanol,3,4-dimethoxy-.beta.-phenethyl alcohol,1-2-hydroxyethyl-3,4-dimethoxybenzene
81911
2-(3,4-dimethoxyphenyl)ethanol
COC1=C(C=C(C=C1)CCO)OC
Color Yellow
Melting Point 44°C to 48°C
Flash Point >110°C
Boiling Point 172°C to 174°C (17.0mmHg)
Infrared Spectrum Authentic
Quantity 50g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula (CH3O)2C6H3CH2CH2OH
Beilstein 06, III, 6333
Formula Weight 182.22
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxyphenethyl alcohol
Show More Show Less

EINECSNumber : 231-032-5