Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxyphenol, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
2033-89-8
C8H10O3
154.17
SMFFZOQLHYIRDA-UHFFFAOYSA-N
phenol, 3,4-dimethoxy,4-hydroxyveratrole,3,4-bis methyloxy phenol,unii-38b43wcu83,3,4-dimethoxy phenol,3,4-dimethoxyphel,3,4,dimethoxyphenol,phenol,4-dimethoxy,3,4-dimethoxy-pheno,pubchem4113
16251
3,4-dimethoxyphenol
COC1=C(C=C(C=C1)O)OC
2033-89-8
C8H10O3
154.17
SMFFZOQLHYIRDA-UHFFFAOYSA-N
phenol, 3,4-dimethoxy,4-hydroxyveratrole,3,4-bis methyloxy phenol,unii-38b43wcu83,3,4-dimethoxy phenol,3,4-dimethoxyphel,3,4,dimethoxyphenol,phenol,4-dimethoxy,3,4-dimethoxy-pheno,pubchem4113
16251
3,4-dimethoxyphenol
COC1=C(C=C(C=C1)O)OC
Infrared Spectrum Authentic
Quantity 10g
Melting Point 77.0°C to 82.0°C
Packaging Glass bottle
Color White to Beige
Assay Percent Range 96% min. (GC)
Linear Formula (CH3O)2C6H3OH
Beilstein 06,III,6277
Formula Weight 154.17
Physical Form Fine Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3, 4-Dimethoxyphenol
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 217-995-4

Recommended Storage : Product may darken during storage