Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxyphenylacetic acid hydrazide, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
60075-23-2
C10H14N2O3
210.23
MFCD00051701
HRMXYTRKEOUMNG-UHFFFAOYSA-N
2-3,4-dimethoxyphenyl acetohydrazide, 3,4-dimethoxyphenylacetic acid hydrazide, 3,4-dimethoxyphenylacethydrazide, 3,4-dimethoxyphenyl acetic hydrazide, acmc-20aneo, homoveratric hydrazide, maybridge1_007491, 2-3,4-dimethoxyphenyl ethanehydrazide, 2-3,4-dimethoxyphenyl ethanohydrazide, 2-3,4-dimethoxyphenyl acetic hydrazide
282482
2-(3,4-dimethoxyphenyl)acetohydrazide
COC1=CC=C(CC(=O)NN)C=C1OC
60075-23-2
C10H14N2O3
210.23
MFCD00051701
HRMXYTRKEOUMNG-UHFFFAOYSA-N
2-3,4-dimethoxyphenyl acetohydrazide, 3,4-dimethoxyphenylacetic acid hydrazide, 3,4-dimethoxyphenylacethydrazide, 3,4-dimethoxyphenyl acetic hydrazide, acmc-20aneo, homoveratric hydrazide, maybridge1_007491, 2-3,4-dimethoxyphenyl ethanehydrazide, 2-3,4-dimethoxyphenyl ethanohydrazide, 2-3,4-dimethoxyphenyl acetic hydrazide
282482
2-(3,4-dimethoxyphenyl)acetohydrazide
COC1=CC=C(CC(=O)NN)C=C1OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 102°C to 104°C
Beilstein 2698478
Quantity 5g
Formula Weight 210.23
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxyphenylacetic acid hydrazide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures