Alfa Aesar™ 3',4'-Dimethylacetophenone, 98%

Manufacturer: Alfa Aesar™ A1218722

Quantity 100g

 View more versions of this product

Catalog No. AAA1218722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3637-01-2
    Molecular Formula C10H12O
    Formula Weight 148.21
    MDL Number MFCD00008743
    Beilstein 907053
    Chemical Name or Material 3',4'-Dimethylacetophenone

    Specifications

    Formula Weight 148.21
    TSCA TSCA
    Boiling Point 242° to 243°C
    MDL Number MFCD00008743
    Beilstein 907053
    Flash Point 102°C (215°F)
    Molecular Formula C10H12O
    CAS 3637-01-2
    Quantity 100g
    Chemical Name or Material 3',4'-Dimethylacetophenone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit