Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethylaniline, 99.5%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
95-64-7
C8H11N
121.18
MFCD00007810
DOLQYFPDPKPQSS-UHFFFAOYSA-N
3,4-xylidine,4-amino-o-xylene,3,4-xylylamine,3,4-dimethylphenylamine,benzenamine, 3,4-dimethyl,3,4-dimethylbenzenamine,3,4-dimethylaminobenzene,1-amino-3,4-dimethylbenzene,4-amino-1,2-dimethylbenzene,aniline, 3,4-dimethyl
7248
CHEBI:39901
3,4-dimethylaniline
CC1=C(C=C(C=C1)N)C
95-64-7
C8H11N
121.18
MFCD00007810
DOLQYFPDPKPQSS-UHFFFAOYSA-N
3,4-xylidine,4-amino-o-xylene,3,4-xylylamine,3,4-dimethylphenylamine,benzenamine, 3,4-dimethyl,3,4-dimethylbenzenamine,3,4-dimethylaminobenzene,1-amino-3,4-dimethylbenzene,4-amino-1,2-dimethylbenzene,aniline, 3,4-dimethyl
7248
CHEBI:39901
3,4-dimethylaniline
CC1=C(C=C(C=C1)N)C
Boiling Point 226°C
Flash Point 98°C
Packaging Glass bottle
Melting Point 49°C to 51°C
Color Beige to Brown
Infrared Spectrum Authentic
Quantity 100g
CAS Min % 99.4
CAS Max % 100.0
Assay Percent Range 99.4% min. (GC)
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1103
Merck Index 15, 10282
Solubility Information Solubility in water: <1g/L (24°C). Other solubilities: slightly soluble in chloroform,soluble in alcohol (>100g/L),diethyl ether,,petroleum ether,very soluble in ligroin
Formula Weight 121.18
Physical Form Crystalline Mass or Crystals
Percent Purity 99.50%
Chemical Name or Material 3, 4-Dimethylaniline, 99.5%
Show More Show Less

GHS H Statement
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 202-437-4

RTECSNumber : ZE9450000

TSCA : TSCA

Recommended Storage : product darkens if exposed to light and air