Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,4-Dimethylaniline, 99.5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 49°C to 51°C |
Color | Beige to Brown |
CAS Min % | 99.4 |
CAS Max % | 100.0 |
Boiling Point | 226°C |
Quantity | 5 g |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Physical Form | Crystalline Mass or Crystals |
Assay Percent Range | 99.4% min. (GC) |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Toxic in contact with skin. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Toxic if inhaled. Harmful if swallowed. |
Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: |
EINECSNumber | 202-437-4 |
RTECSNumber | ZE9450000 |
TSCA | TSCA |
Recommended Storage | product darkens if exposed to light and air |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction