Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3,4-Dimethylbenzenesulfonyl chloride, 98%
Click to view available options
Quantity:
5g
25g
100g
Description
Applications
3,4-Dimethylbenzenesulfonyl chloride is used as an intermediate.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agent, bases, water/ moisture.
3,4-Dimethylbenzenesulfonyl chloride is used as an intermediate.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agent, bases, water/ moisture.
Specifications
Quantity | 25g |
Odor | Acrid |
UN Number | UN3261 |
Solubility Information | Reacts with water. |
Formula Weight | 204.68 |
Percent Purity | 98% |
Sensitivity | Moisture sensitive |
Chemical Name or Material | 3,4-Dimethylbenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 210-337-7
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen