Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethylbenzyl chloride, 97%, Thermo Scientific™

Catalog Number ac436890250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
102-46-5
C9H11Cl
154.64
UBQRAAXAHIKWRI-UHFFFAOYSA-N
3,4-dimethylbenzyl chloride, 4-chloromethyl-1,2-dimethylbenzene, benzene, 4-chloromethyl-1,2-dimethyl, benzene, 1-chloromethyl-3,4-dimethyl, 4-chloromethyl-o-xylene, 4-chloromethyl-1,2-dimethyl-benzene, 1-chloromethyl-3,4-dimethylbenzene, acmc-20anej, pubchem14890
66018
4-(chloromethyl)-1,2-dimethylbenzene
CC1=C(C=C(C=C1)CCl)C
102-46-5
C9H11Cl
154.64
UBQRAAXAHIKWRI-UHFFFAOYSA-N
3,4-dimethylbenzyl chloride, 4-chloromethyl-1,2-dimethylbenzene, benzene, 4-chloromethyl-1,2-dimethyl, benzene, 1-chloromethyl-3,4-dimethyl, 4-chloromethyl-o-xylene, 4-chloromethyl-1,2-dimethyl-benzene, 1-chloromethyl-3,4-dimethylbenzene, acmc-20anej, pubchem14890
66018
4-(chloromethyl)-1,2-dimethylbenzene
CC1=C(C=C(C=C1)CCl)C
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 98°C (12.0 Torr)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 154.64
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 4-Dimethylbenzyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
EINECSNumber 203-032-5

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit