Alfa Aesar™ 3,4'-Dimethyldiphenyl ether, 97%

Manufacturer: Alfa Aesar™ B2267809

Quantity 10g

 View more versions of this product

Catalog No. AAB2267809

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 51801-69-5
    Molecular Formula C14H14O
    Formula Weight 198.26
    MDL Number MFCD00151815
    Beilstein 1951603
    Chemical Name or Material 3,4'-Dimethyldiphenyl ether

    Specifications

    Formula Weight 198.26
    Boiling Point 164°C (10mmHg)
    MDL Number MFCD00151815
    Quantity 10g
    Molecular Formula C14H14O
    CAS 51801-69-5
    Beilstein 1951603
    Chemical Name or Material 3,4'-Dimethyldiphenyl ether
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit