Alfa Aesar™ 3,4-Dimethylphenol, 98+%

Manufacturer: Alfa Aesar™ A1059936

Quantity 500g

 View more versions of this product

Catalog No. AAA1059936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 95-65-8
    Molecular Formula C8H10O
    Formula Weight 122.17
    MDL Number MFCD00002304
    UN Number UN2261
    Beilstein 1099267
    Merck Index 14,10082
    Synonym 3,4-Xylenol
    Chemical Name or Material 3,4-Dimethylphenol

    Specifications

    Formula Weight 122.17
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym 3,4-Xylenol
    Boiling Point 227°C
    Melting Point 65° to 68°C
    Quantity 500g
    TSCA TSCA
    MDL Number MFCD00002304
    CAS 95-65-8
    UN Number UN2261
    Beilstein 1099267
    Health Hazard 2 GHS H Statement
    H310-H314-H318-H302
    Fatal in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID
    Molecular Formula C8H10O
    Merck Index 14,10082
    Health Hazard 1 Danger
    RTECS Number ZE6300000
    Chemical Name or Material 3,4-Dimethylphenol
    Assay ≥98%

    Description

    Applications
    3,4-Dimethylphenol, is used as an anti-Infective agents. It has wide applications in the analytical techniques such as Gas Chromatography, Reversed Phase Chromatography, HPLC and UHPLC.

    Solubility
    Slightly soluble in water. Less than 1 mg/mL at 68° F.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Light sensitive.Hygroscopic. Incompatible with bases, acid chlorides, acid anhydrides, and oxidizing agents.

    GHS H Statement
    H310-H314-H318-H302
    Fatal in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit