Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dimethylphenol, 98+%

Manufacturer:  Alfa Aesar™ A1059922

Quantity 100g

 View more versions of this product

Catalog No. AAA1059922


  Check Availability
Add to cart

Specifications

Description

Applications
3,4-Dimethylphenol, is used as an anti-Infective agents. It has wide applications in the analytical techniques such as Gas Chromatography, Reversed Phase Chromatography, HPLC and UHPLC.

Solubility
Slightly soluble in water. Less than 1 mg/mL at 68° F.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Light sensitive.Hygroscopic. Incompatible with bases, acid chlorides, acid anhydrides, and oxidizing agents.
Description & Specifications

Chemical Identifiers

95-65-8
122.17
UN2261
14,10082
3,4-Dimethylphenol
C8H10O
MFCD00002304
1099267
3,4-Xylenol

Specifications

227°C
100g
C8H10O
MFCD00002304
1099267
3,4-Xylenol
≥98%
65° to 68°C
95-65-8
122.17
UN2261
14,10082
3,4-Dimethylphenol

Safety and Handling

SDS & Safety Documents

GHS H Statement
H310-H314-H318-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID

RTECSNumber : ZE6300000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit