Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethylphenol 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
95-65-8
C8H10O
122.17
MFCD00002304
YCOXTKKNXUZSKD-UHFFFAOYSA-N
3,4-xylenol,phenol, 3,4-dimethyl,4,5-dimethylphenol,3,4-dimethyl phenol,1,3,4-xylenol,4-hydroxy-1,2-dimethylbenzene,1-hydroxy-3,4-dimethylbenzene,3,4-dimethyl-phenol,3,4-dmp,ccris 723
7249
CHEBI:39839
3,4-dimethylphenol
CC1=C(C=C(C=C1)O)C
95-65-8
C8H10O
122.17
MFCD00002304
YCOXTKKNXUZSKD-UHFFFAOYSA-N
3,4-xylenol,phenol, 3,4-dimethyl,4,5-dimethylphenol,3,4-dimethyl phenol,1,3,4-xylenol,4-hydroxy-1,2-dimethylbenzene,1-hydroxy-3,4-dimethylbenzene,3,4-dimethyl-phenol,3,4-dmp,ccris 723
7249
CHEBI:39839
3,4-dimethylphenol
CC1=C(C=C(C=C1)O)C
Boiling Point 227°C
Density 0.9830g/mL
Flash Point 80°C
Specific Gravity 0.983
Color Cream to White
Quantity 100g
Packaging Glass bottle
Melting Point 63°C to 67°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2C6H3OH
Beilstein 06, 480
Merck Index 15, 10279
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in naoh solutions,freely soluble in alcohol,chloroform,ether and,benzene
Formula Weight 122.17
Physical Form Crystalline Powder
Percent Purity ≥98.5% (GC)
Chemical Name or Material 3, 4-Dimethylphenol, 99%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-439-5

RTECSNumber : ZE6300000

TSCA : TSCA