Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dimethylphenoxyacetic acid, 98%

Click to view available options
Quantity:
5g
25g
13335-73-4
C10H12O3
180.203
MFCD00017595
GIEQJKZWTIFEJM-UHFFFAOYSA-N
2-3,4-dimethylphenoxy acetic acid,3,4-dimethylphenoxyacetic acid,3,4-dimethylphenoxy acetic acid,3,4-xylyloxyacetic acid,acmc-1bqwp,3,4-dimethyl-phenoxy acetic acid,3,4-dimethyl-phenoxy-acetic acid,acetic acid,2-3,4-dimethylphenoxy,acetic acid, 2-3,4-dimethylphenoxy
83355
2-(3,4-dimethylphenoxy)acetic acid
CC1=C(C=C(C=C1)OCC(=O)O)C
13335-73-4
C10H12O3
180.203
MFCD00017595
GIEQJKZWTIFEJM-UHFFFAOYSA-N
2-3,4-dimethylphenoxy acetic acid,3,4-dimethylphenoxyacetic acid,3,4-dimethylphenoxy acetic acid,3,4-xylyloxyacetic acid,acmc-1bqwp,3,4-dimethyl-phenoxy acetic acid,3,4-dimethyl-phenoxy-acetic acid,acetic acid,2-3,4-dimethylphenoxy,acetic acid, 2-3,4-dimethylphenoxy
83355
2-(3,4-dimethylphenoxy)acetic acid
CC1=C(C=C(C=C1)OCC(=O)O)C
Melting Point 160°C to 165°C
Quantity 25g
Beilstein 2558866
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylphenoxyacetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 236-386-4

TSCA : No

Recommended Storage : Ambient temperatures