Alfa Aesar™ 3,4-Dimethylthiophenol, 98%

Manufacturer: Alfa Aesar™ B2316814

Quantity 25g

 View more versions of this product

Catalog No. AAB2316814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18800-53-8
    Molecular Formula C8H10S
    Formula Weight 138.23
    MDL Number MFCD00010023
    UN Number UN2810
    Beilstein 2040353
    Synonym 3,4-Dimethylbenzenethiol
    Chemical Name or Material 3,4-Dimethylthiophenol

    Specifications

    Formula Weight 138.23
    TSCA TSCA
    Boiling Point 218°C
    Quantity 25g
    Flash Point 90°C (194°F)
    Odor Unpleasant
    UN Number UN2810
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319-H335-H227
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Molecular Formula C8H10S
    MDL Number MFCD00010023
    CAS 18800-53-8
    Beilstein 2040353
    Synonym 3,4-Dimethylbenzenethiol
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P260-P280h-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Chemical Name or Material 3,4-Dimethylthiophenol
    Assay 98%

    GHS H Statement
    H302-H312-H332-H315-H319-H335-H227
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P260-P280h-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit