Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(4-Fluorophenyl)-1H-pyrazole, 97%

Manufacturer:  Alfa Aesar™ H3435509

Quantity 10g

 View more versions of this product

Catalog No. AAH3435509


  Check Availability
Add to cart

Specifications

Description

Applications
3-(4-Fluorophenyl)-1H-pyrazole is used as pharmaceutical intermediates.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

154258-82-9
162.17
3-(4-Fluorophenyl)-1H-pyrazole
C9H7FN2
MFCD00665857

Specifications

101° to 103°C
154258-82-9
162.17
3-(4-Fluorophenyl)-1H-pyrazole
10g
C9H7FN2
MFCD00665857
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit