Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Fluorophenyl)propionic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
459-31-4
C9H9FO2
168.167
MFCD00060327
ZMKXWDPUXLPHCA-UHFFFAOYSA-N
3-4-fluorophenyl propionic acid,3-4-fluorophenyl propanoic acid,benzenepropanoic acid, 4-fluoro,3-4-fluoro-phenyl-propionic acid,3-4-fluorophenyl propionicacid,3-4'-fluorophenyl propanoic acid,acmc-1aehu,4-fluorohydrocinnamic acid,4-fluorophenylpropionic acid,4-fluorobenzenepropanoic acid
136302
3-(4-fluorophenyl)propanoic acid
C1=CC(=CC=C1CCC(=O)O)F
459-31-4
C9H9FO2
168.167
MFCD00060327
ZMKXWDPUXLPHCA-UHFFFAOYSA-N
3-4-fluorophenyl propionic acid,3-4-fluorophenyl propanoic acid,benzenepropanoic acid, 4-fluoro,3-4-fluoro-phenyl-propionic acid,3-4-fluorophenyl propionicacid,3-4'-fluorophenyl propanoic acid,acmc-1aehu,4-fluorohydrocinnamic acid,4-fluorophenylpropionic acid,4-fluorobenzenepropanoic acid
136302
3-(4-fluorophenyl)propanoic acid
C1=CC(=CC=C1CCC(=O)O)F
Melting Point 86°C to 91°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula COOH(CH2)2C6H4F
Formula Weight 168.17
Physical Form Crystalline Powder or Crystals
Percent Purity ≥97.5%
Chemical Name or Material 3-(4-Fluorophenyl)propionic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.