Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Isopropylidene-D-mannitol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
3969-84-4
C9H18O6
222.237
MFCD00075122
YCOMFYACDCWMMD-WCTZXXKLSA-N
3,4-o-isopropylidene-d-mannitol,1r,1'r-1,1'-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl bis ethane-1,2-diol,1r-1-4r,5r-5-1r-1,2-dihydroxyethyl-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol,oi4-o-isopropylidene-d-mannitol,d-mannitol-3,4-acetonide,3-o,4-o-isopropylidene-d-mannitol,3,4-isopropylidene-d-mannitol,3,4-o-1-methylethylidene-d-mannitol,1r,1'r-1,1'-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl diethane-1,2-diol
7096260
(1R)-1-[(4R,5R)-5-[(1R)-1,2-dihydroxyethyl]-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol
CC1(OC(C(O1)C(CO)O)C(CO)O)C
3969-84-4
C9H18O6
222.237
MFCD00075122
YCOMFYACDCWMMD-WCTZXXKLSA-N
3,4-o-isopropylidene-d-mannitol,1r,1'r-1,1'-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl bis ethane-1,2-diol,1r-1-4r,5r-5-1r-1,2-dihydroxyethyl-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol,oi4-o-isopropylidene-d-mannitol,d-mannitol-3,4-acetonide,3-o,4-o-isopropylidene-d-mannitol,3,4-isopropylidene-d-mannitol,3,4-o-1-methylethylidene-d-mannitol,1r,1'r-1,1'-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl diethane-1,2-diol
7096260
(1R)-1-[(4R,5R)-5-[(1R)-1,2-dihydroxyethyl]-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol
CC1(OC(C(O1)C(CO)O)C(CO)O)C
Quantity 5g
Color White
Specific Rotation +29° to +31° (20°C, 589nm) (c=3, H2O)
Infrared Spectrum Authentic
Specific Rotation Condition + 30.00 (20.00°C c=3,H2O)
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 222.24
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 3, 4-Isopropylidene-D-mannitol, 98%
Show More Show Less