Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Methoxyphenoxy)benzaldehyde, 95%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
62373-80-2
C14H12O3
228.247
MFCD00003358
WLFDEVVCXPTAQA-UHFFFAOYSA-N
3-4-methoxyphenoxy benzaldehyde,benzaldehyde, 3-4-methoxyphenoxy,acmc-209n2y,3-4-methoxyphenoxy-benzaldehyde,3-formyl-4'-methoxydiphenyl ether,3-4-methoxy phenoxy benzaldehyde
123553
3-(4-methoxyphenoxy)benzaldehyde
COC1=CC=C(C=C1)OC2=CC=CC(=C2)C=O
62373-80-2
C14H12O3
228.247
MFCD00003358
WLFDEVVCXPTAQA-UHFFFAOYSA-N
3-4-methoxyphenoxy benzaldehyde,benzaldehyde, 3-4-methoxyphenoxy,acmc-209n2y,3-4-methoxyphenoxy-benzaldehyde,3-formyl-4'-methoxydiphenyl ether,3-4-methoxy phenoxy benzaldehyde
123553
3-(4-methoxyphenoxy)benzaldehyde
COC1=CC=C(C=C1)OC2=CC=CC(=C2)C=O
Infrared Spectrum Authentic
Quantity 10g
Color Yellow
Density 1.0800g/mL
Packaging Glass bottle
Refractive Index 1.5920 to 1.5970 (20°C, 589nm)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula CH3OC6H4OC6H4CHO
Formula Weight 228.25
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material 3-(4-Methoxyphenoxy)benzaldehyde, 95%
Show More Show Less

EINECSNumber : 263-527-7

TSCA : TSCA