Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Methoxyphenyl)pyrazole, 98%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
27069-17-6
C10H10N2O
174.2
MFCD00112008
QUSABYOAMXPMQH-UHFFFAOYSA-N
3-4-methoxyphenyl-1h-pyrazole,3-4-methoxyphenyl pyrazole,5-4-methoxyphenyl-1h-pyrazole,chembl39210,3-4-methoxy-phenyl-1h-pyrazole,1h-pyrazole, 3-4-methoxyphenyl,4-methoxy-1-pyrazol-3-ylbenzene,3-4-methoxyphenyl-2h-pyrazole,maybridge1_007801,3-4-methoxyphenyl-pyrazole
599984
5-(4-methoxyphenyl)-1H-pyrazole
COC1=CC=C(C=C1)C2=CC=NN2
27069-17-6
C10H10N2O
174.2
MFCD00112008
QUSABYOAMXPMQH-UHFFFAOYSA-N
3-4-methoxyphenyl-1h-pyrazole,3-4-methoxyphenyl pyrazole,5-4-methoxyphenyl-1h-pyrazole,chembl39210,3-4-methoxy-phenyl-1h-pyrazole,1h-pyrazole, 3-4-methoxyphenyl,4-methoxy-1-pyrazol-3-ylbenzene,3-4-methoxyphenyl-2h-pyrazole,maybridge1_007801,3-4-methoxyphenyl-pyrazole
599984
5-(4-methoxyphenyl)-1H-pyrazole
COC1=CC=C(C=C1)C2=CC=NN2
Infrared Spectrum Authentic
Color White to Yellow
Boiling Point 185°C (0.6mmHg)
Melting Point 127°C to 130°C
Quantity 1g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 174.2
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 3-(4-Methoxyphenyl)pyrazole
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning