Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Methylbenzoyl)propionic acid, 98%, Acros Organics

Click to view available options
Quantity:
5g
4619-20-9
C11H12O3
192.214
MFCD00020541
OEEUWZITKKSXAZ-UHFFFAOYSA-N
3-4-methylbenzoyl propionic acid,4-oxo-4-p-tolyl butanoic acid,4-4-methylphenyl-4-oxobutanoic acid,4-4-methylphenyl-4-oxobutyric acid,4-oxo-4-4-tolyl butyric acid,4-oxo-4-p-tolylbutyric acid,4-oxo-4-p-tolylbutanoic acid,3-4-methylbenzoyl-propionic acid,acmc-1aooh,benzenebutanoic acid, 4-methyl-gamma-oxo
244162
4-(4-methylphenyl)-4-oxobutanoic acid
CC1=CC=C(C=C1)C(=O)CCC(=O)O
4619-20-9
C11H12O3
192.214
MFCD00020541
OEEUWZITKKSXAZ-UHFFFAOYSA-N
3-4-methylbenzoyl propionic acid,4-oxo-4-p-tolyl butanoic acid,4-4-methylphenyl-4-oxobutanoic acid,4-4-methylphenyl-4-oxobutyric acid,4-oxo-4-4-tolyl butyric acid,4-oxo-4-p-tolylbutyric acid,4-oxo-4-p-tolylbutanoic acid,3-4-methylbenzoyl-propionic acid,acmc-1aooh,benzenebutanoic acid, 4-methyl-gamma-oxo
244162
4-(4-methylphenyl)-4-oxobutanoic acid
CC1=CC=C(C=C1)C(=O)CCC(=O)O
Quantity 5g
Melting Point 127°C to 130°C
Color White to Beige
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H4COCHCHCOOH
Formula Weight 192.21
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-(4-Methylbenzoyl)propionic acid, 98%
Show More Show Less