Alfa Aesar™ 3,4-(Methylenedioxy)benzylamine, 97%

Manufacturer: Alfa Aesar™ B2183714

Quantity 25g

 View more versions of this product

Catalog No. AAB2183714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2620-50-0
    Molecular Formula C8H9NO2
    Formula Weight 151.17
    MDL Number MFCD00005840
    UN Number UN2735
    Synonym (1,3-Benzodioxol-5-yl)methylamine; Piperonylamine
    Chemical Name or Material 3,4-(Methylenedioxy)benzylamine

    Specifications

    Formula Weight 151.17
    Molecular Formula C8H9NO2
    Quantity 25g
    Flash Point >110°C (230°F)
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    MDL Number MFCD00005840
    CAS 2620-50-0
    UN Number UN2735
    Synonym (1,3-Benzodioxol-5-yl)methylamine; Piperonylamine
    Health Hazard 1 Danger
    Boiling Point 136° to 137°C (11mmHg)
    Odor Amine-like
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    Chemical Name or Material 3,4-(Methylenedioxy)benzylamine
    Assay 97%

    Description

    Applications
    Piperonylamine is a metabolite of dopamine analogs. It is a pharmaceutical intermediate.

    Solubility
    Sparingly soluble in water. (0.9 g/100 mL).

    Notes
    Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit