Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-(Methylenedioxy)benzylamine, 97%

Click to view available options
Quantity:
5g
25g
100g
2620-50-0
C8H9NO2
151.165
MFCD00005840
ZILSBZLQGRBMOR-UHFFFAOYSA-N
piperonylamine,benzo d 1,3 dioxol-5-ylmethanamine,3,4-methylenedioxybenzylamine,1,3-benzodioxol-5-ylmethylamine,1,3-benzodioxole-5-methanamine,2h-1,3-benzodioxol-5-ylmethanamine,3,4-methylenedioxy benzylamine,benzo-1,3-dioxole-5-methylamine,1-1,3-benzodioxol-5-yl methanamine,5-aminomethyl-1,3-benzodioxole
75799
1,3-benzodioxol-5-ylmethanamine
C1OC2=C(O1)C=C(C=C2)CN
2620-50-0
C8H9NO2
151.165
MFCD00005840
ZILSBZLQGRBMOR-UHFFFAOYSA-N
piperonylamine,benzo d 1,3 dioxol-5-ylmethanamine,3,4-methylenedioxybenzylamine,1,3-benzodioxol-5-ylmethylamine,1,3-benzodioxole-5-methanamine,2h-1,3-benzodioxol-5-ylmethanamine,3,4-methylenedioxy benzylamine,benzo-1,3-dioxole-5-methylamine,1-1,3-benzodioxol-5-yl methanamine,5-aminomethyl-1,3-benzodioxole
75799
1,3-benzodioxol-5-ylmethanamine
C1OC2=C(O1)C=C(C=C2)CN
Applications
Piperonylamine is a metabolite of dopamine analogs. It is a pharmaceutical intermediate.

Solubility
Sparingly soluble in water. (0.9 g/100 mL).

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.
Flash Point >110°C (230°F)
Odor Amine-like
Boiling Point 136°C to 137°C (11mmHg)
Quantity 100g
UN Number UN2735
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material 3,4-(Methylenedioxy)benzylamine

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.