Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-(Methylenedioxy)cinnamic acid, predominantly trans, 99%, Thermo Scientific Chemicals

Catalog Number AAA1012214
Click to view available options
Quantity:
25 g
100 g
500 g
2373-80-0
C10H8O4
192.17
MFCD00005837
QFQYZMGOKIROEC-DUXPYHPUSA-N
3,4-methylenedioxycinnamic acid, 3,4-methylenedioxy cinnamic acid, 3-benzo d 1,3 dioxol-5-yl acrylic acid, unii-x8h0wpj08y, 3-1,3-benzodioxol-5-yl acrylic acid, 2e-3-2h-1,3-benzodioxol-5-yl prop-2-enoic acid, methylenedioxycinnamic acid, x8h0wpj08y, 2e-3-1,3-benzodioxol-5-yl acrylic acid, 2e-3-1,3-benzodioxol-5-yl-2-propenoic acid
643181
CHEBI:81482
(E)-3-(1,3-benzodioxol-5-yl)prop-2-enoic acid
C1OC2=C(O1)C=C(C=C2)C=CC(=O)O
2373-80-0
C10H8O4
192.17
MFCD00005837
QFQYZMGOKIROEC-DUXPYHPUSA-N
3,4-methylenedioxycinnamic acid, 3,4-methylenedioxy cinnamic acid, 3-benzo d 1,3 dioxol-5-yl acrylic acid, unii-x8h0wpj08y, 3-1,3-benzodioxol-5-yl acrylic acid, 2e-3-2h-1,3-benzodioxol-5-yl prop-2-enoic acid, methylenedioxycinnamic acid, x8h0wpj08y, 2e-3-1,3-benzodioxol-5-yl acrylic acid, 2e-3-1,3-benzodioxol-5-yl-2-propenoic acid
643181
CHEBI:81482
(E)-3-(1,3-benzodioxol-5-yl)prop-2-enoic acid
C1OC2=C(O1)C=C(C=C2)C=CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼243°C (decomposition)
Quantity 25 g
Beilstein 84599
Formula Weight 192.17
Percent Purity 0.99
Chemical Name or Material 3,4- (Methylenedioxy)cinnamic acid, predominantly trans
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 219-151-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit