Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Methylenedioxyaniline 98.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
500g
14268-66-7
C7H7NO2
137.138
MFCD00005832
XGNXYCFREOZBOL-UHFFFAOYSA-N
3,4-methylenedioxy aniline,benzo d 1,3 dioxol-5-amine,5-amino-1,3-benzodioxole,3,4-methylenedioxyaniline,4-amino-1,2-methylenedioxybenzene,5-aminobenzodioxole,methylenedioxyaniline,2h-1,3-benzodioxol-5-amine,1-amino-3,4-methylenedioxybenzene,1-amino-3,4-dimethylenedioxybenzene
84310
1,3-benzodioxol-5-amine
C1OC2=C(O1)C=C(C=C2)N
14268-66-7
C7H7NO2
137.138
MFCD00005832
XGNXYCFREOZBOL-UHFFFAOYSA-N
3,4-methylenedioxy aniline,benzo d 1,3 dioxol-5-amine,5-amino-1,3-benzodioxole,3,4-methylenedioxyaniline,4-amino-1,2-methylenedioxybenzene,5-aminobenzodioxole,methylenedioxyaniline,2h-1,3-benzodioxol-5-amine,1-amino-3,4-methylenedioxybenzene,1-amino-3,4-dimethylenedioxybenzene
84310
1,3-benzodioxol-5-amine
C1OC2=C(O1)C=C(C=C2)N
Boiling Point 156°C
Color Undesignated
Melting Point 44°C
Quantity 25g
Formula Weight 137.14
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 3,4-Methylenedioxyaniline

TSCA : Yes

Recommended Storage : Refrigerator