Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Methylenedioxyphenylacetonitrile 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
4439-02-5
C9H7NO2
161.16
MFCD00005835
ZQPBOYASBNAXOZ-UHFFFAOYSA-N
3,4-methylenedioxy phenylacetonitrile, 1,3-benzodioxole-5-acetonitrile, 3,4-methylenedioxyphenylacetonitrile, 2-benzo d 1,3 dioxol-5-yl acetonitrile, 3,4-methylenedioxybenzyl cyanide, 2-2h-1,3-benzodioxol-5-yl acetonitrile, 1,3-benzodioxol-5-ylacetonitrile, 2-1,3-benzodioxol-5-yl acetonitrile, 3,4-methylenedioxybenzylcyanide, homopiperonylonitrile
78178
2-(1,3-benzodioxol-5-yl)acetonitrile
C1OC2=C(O1)C=C(C=C2)CC#N
4439-02-5
C9H7NO2
161.16
MFCD00005835
ZQPBOYASBNAXOZ-UHFFFAOYSA-N
3,4-methylenedioxy phenylacetonitrile, 1,3-benzodioxole-5-acetonitrile, 3,4-methylenedioxyphenylacetonitrile, 2-benzo d 1,3 dioxol-5-yl acetonitrile, 3,4-methylenedioxybenzyl cyanide, 2-2h-1,3-benzodioxol-5-yl acetonitrile, 1,3-benzodioxol-5-ylacetonitrile, 2-1,3-benzodioxol-5-yl acetonitrile, 3,4-methylenedioxybenzylcyanide, homopiperonylonitrile
78178
2-(1,3-benzodioxol-5-yl)acetonitrile
C1OC2=C(O1)C=C(C=C2)CC#N
Color White-Yellow
Melting Point 43°C
Boiling Point 140°C
Quantity 25g
Formula Weight 161.16
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3,4-Methylenedioxyphenylacetonitrile

EINECSNumber : (5)-3564

TSCA : No