Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(4-Morpholinyl)propiophenone hydrochloride, 98+%

Click to view available options
Quantity:
5g
25g
100g
1020-16-2
C13H18ClNO2
255.742
MFCD00035332
BEUAFINDWSQCGS-UHFFFAOYSA-N
3-morpholinopropiophenone hydrochloride,3-4-morpholino propiophenone hydrochloride,3-4-morpholinyl propiophenone hydrochloride,3-morpholino-1-phenylpropan-1-one hydrochloride,labotest-bb lt00454997,3-4-morpholinyl-1-phenyl-1-propanone hydrochloride,3-morpholin-4-yl-1-phenylpropan-1-one hydrochloride,3-morpholinopropiophenone hcl
3083670
3-morpholin-4-yl-1-phenylpropan-1-one;hydrochloride
C1COCCN1CCC(=O)C2=CC=CC=C2.Cl
1020-16-2
C13H18ClNO2
255.742
MFCD00035332
BEUAFINDWSQCGS-UHFFFAOYSA-N
3-morpholinopropiophenone hydrochloride,3-4-morpholino propiophenone hydrochloride,3-4-morpholinyl propiophenone hydrochloride,3-morpholino-1-phenylpropan-1-one hydrochloride,labotest-bb lt00454997,3-4-morpholinyl-1-phenyl-1-propanone hydrochloride,3-morpholin-4-yl-1-phenylpropan-1-one hydrochloride,3-morpholinopropiophenone hcl
3083670
3-morpholin-4-yl-1-phenylpropan-1-one;hydrochloride
C1COCCN1CCC(=O)C2=CC=CC=C2.Cl
Melting Point 180°C to 182°C
Quantity 25g
Beilstein 3723608
Formula Weight 255.76
Percent Purity ≥98%
Chemical Name or Material 3-(4-Morpholinyl)propiophenone hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning