Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-(4-Morpholinyl)propylamine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Fully miscible in water.
Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with acids, acid chlorides, acid anhydrides.
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Fully miscible in water.
Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with acids, acid chlorides, acid anhydrides.
Specifications
Melting Point | -15°C |
Density | 0.987 |
Boiling Point | 223°C to 225°C |
Flash Point | 98°C (210°F) |
Refractive Index | 1.4761 |
Quantity | 50 g |
UN Number | UN2735 |
Sensitivity | Air Sensitive |
Solubility Information | Fully miscible in water. |
Formula Weight | 144.22 |
Show More |
Safety and Handling
Hazard Category | H302+H312-H314 |
Hazard Statement | GHS H Statement H311-H314-H318 Toxic in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S. |
EINECSNumber | 204-590-2 |
RTECSNumber | QD7700000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction