Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(4-Morpholinyl)propylamine, 98%

Click to view available options
Quantity:
50g
250g
123-00-2
C7H16N2O
144.218
MFCD00006184
UIKUBYKUYUSRSM-UHFFFAOYSA-N
n-3-aminopropyl morpholine,4-morpholinepropanamine,3-morpholinopropylamine,3-morpholinopropan-1-amine,n-aminopropylmorpholine,4-3-aminopropyl morpholine,4-morpholinepropylamine,4-aminopropylmorpholine,1-amino-3-morpholinopropane,3-4-morpholinyl-1-propanamine
61055
CHEBI:86554
3-morpholin-4-ylpropan-1-amine
C1COCCN1CCCN
123-00-2
C7H16N2O
144.218
MFCD00006184
UIKUBYKUYUSRSM-UHFFFAOYSA-N
n-3-aminopropyl morpholine,4-morpholinepropanamine,3-morpholinopropylamine,3-morpholinopropan-1-amine,n-aminopropylmorpholine,4-3-aminopropyl morpholine,4-morpholinepropylamine,4-aminopropylmorpholine,1-amino-3-morpholinopropane,3-4-morpholinyl-1-propanamine
61055
CHEBI:86554
3-morpholin-4-ylpropan-1-amine
C1COCCN1CCCN
Applications
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with acids, acid chlorides, acid anhydrides.
Flash Point 98°C (210°F)
Boiling Point 223°C to 225°C
Quantity 250g
Melting Point -15°C
UN Number UN2735
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material 3-(4-Morpholinyl)propylamine

GHS H Statement
H311-H314-H318
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

RTECSNumber : QD7700000

TSCA : TSCA