Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(4-Morpholinylmethyl)indole, 95%

Click to view available options
Quantity:
250mg
1g
5g
5379-88-4
C13H16N2O
216.284
MFCD00458413
VHGISLOQNDQFIW-UHFFFAOYSA-N
3-morpholinomethyl indole,indole, 3-morpholinomethyl,3-morpholin-4-ylmethyl-1h-indole,4-27-00-00551 beilstein handbook reference,4-indol-3-ylmethyl morpholine,3-morpholinomethyl-1h-indole,morpholine, n-3-indolyl methyl,4-1h-indol-3-ylmethyl morpholine,3-4-morpholinylmethyl indole
21477
4-(1H-indol-3-ylmethyl)morpholine
C1COCCN1CC2=CNC3=CC=CC=C32
5379-88-4
C13H16N2O
216.284
MFCD00458413
VHGISLOQNDQFIW-UHFFFAOYSA-N
3-morpholinomethyl indole,indole, 3-morpholinomethyl,3-morpholin-4-ylmethyl-1h-indole,4-27-00-00551 beilstein handbook reference,4-indol-3-ylmethyl morpholine,3-morpholinomethyl-1h-indole,morpholine, n-3-indolyl methyl,4-1h-indol-3-ylmethyl morpholine,3-4-morpholinylmethyl indole
21477
4-(1H-indol-3-ylmethyl)morpholine
C1COCCN1CC2=CNC3=CC=CC=C32
Quantity 250mg
Color White
Beilstein 193005
Formula Weight 216.28
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 3-(4-Morpholinylmethyl)indole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : NM1140000

TSCA : No

Recommended Storage : Ambient temperatures