Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Nitrophenyl)propanoic acid, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
16642-79-8
C9H9NO4
195.18
VZOPVJNBOQOLPN-UHFFFAOYSA-N
3-4-nitrophenyl propanoic acid,3-4-nitrophenyl propionic acid,benzenepropanoic acid, 4-nitro,3-4-nitrophenyl-propionic acid,4-nitrobenzenepropanoic acid,p-nitrohydrozimtsaure,pubchem13615,p-nitrohydrocinnamic acid
85526
C1=CC(=CC=C1CCC(=O)O)[N+](=O)[O-]
16642-79-8
C9H9NO4
195.18
VZOPVJNBOQOLPN-UHFFFAOYSA-N
3-4-nitrophenyl propanoic acid,3-4-nitrophenyl propionic acid,benzenepropanoic acid, 4-nitro,3-4-nitrophenyl-propionic acid,4-nitrobenzenepropanoic acid,p-nitrohydrozimtsaure,pubchem13615,p-nitrohydrocinnamic acid
85526
C1=CC(=CC=C1CCC(=O)O)[N+](=O)[O-]
Infrared Spectrum Authentic
Quantity 25g
Melting Point 166.0°C to 168.0°C
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 195.18
Percent Purity 98%
Chemical Name or Material 3-(4-Nitrophenyl)propanoic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning