Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Nitrophenyl)propanoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7077722
Click to view available options
Quantity:
25 g
100 g
16642-79-8
C9H9NO4
195.17
MFCD00126834
VZOPVJNBOQOLPN-UHFFFAOYSA-N
3-4-nitrophenyl propanoic acid, 3-4-nitrophenyl propionic acid, benzenepropanoic acid, 4-nitro, 3-4-nitrophenyl-propionic acid, 4-nitrobenzenepropanoic acid, p-nitrohydrozimtsaure, pubchem13615, p-nitrohydrocinnamic acid
85526
3-(4-nitrophenyl)propanoic acid
OC(=O)CCC1=CC=C(C=C1)[N+]([O-])=O
16642-79-8
C9H9NO4
195.17
MFCD00126834
VZOPVJNBOQOLPN-UHFFFAOYSA-N
3-4-nitrophenyl propanoic acid, 3-4-nitrophenyl propionic acid, benzenepropanoic acid, 4-nitro, 3-4-nitrophenyl-propionic acid, 4-nitrobenzenepropanoic acid, p-nitrohydrozimtsaure, pubchem13615, p-nitrohydrocinnamic acid
85526
3-(4-nitrophenyl)propanoic acid
OC(=O)CCC1=CC=C(C=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 166.0°C to 168.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 195.18
Percent Purity 98%
Chemical Name or Material 3-(4-Nitrophenyl)propanoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit