Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Pyridinedicarboxylic Acid 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
490-11-9
C7H5NO4
167.12
MFCD00006392
MUYSADWCWFFZKR-UHFFFAOYSA-N
3,4-pyridinedicarboxylic acid,cinchomeronic acid,chinchomeronic acid,3,4-pyridinedicarboxylicacid,pyridine-3,4-dicarboxylicacid,unii-7389uzw5dy,3,4-pyridindicarbons,cinchomeronsaure,pubchem2419,3,4-dicarboxypyridine
10273
CHEBI:46860
pyridine-3,4-dicarboxylic acid
C1=CN=CC(=C1C(=O)O)C(=O)O
490-11-9
C7H5NO4
167.12
MFCD00006392
MUYSADWCWFFZKR-UHFFFAOYSA-N
3,4-pyridinedicarboxylic acid,cinchomeronic acid,chinchomeronic acid,3,4-pyridinedicarboxylicacid,pyridine-3,4-dicarboxylicacid,unii-7389uzw5dy,3,4-pyridindicarbons,cinchomeronsaure,pubchem2419,3,4-dicarboxypyridine
10273
CHEBI:46860
pyridine-3,4-dicarboxylic acid
C1=CN=CC(=C1C(=O)O)C(=O)O
Melting Point 262°C
Color White to Beige
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 22, 155
Merck Index 15, 2286
Solubility Information Solubility in water: .. Other solubilities: sparingly soluble in hot alcohol,ether and benzen,insoluble in chloroform
Formula Weight 167.12
Physical Form Fine Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 3, 4-Pyridinedicarboxylic acid, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 207-705-4

TSCA : TSCA