Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]-propanol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
51268-87-2
C8H16O3
160.21
MFCD01631673
BSLDYXOPGSOQGZ-ZETCQYMHSA-N
3-4s-2,2-dimethyl-1,3-dioxolan-4-yl propan-1-ol,1,3-dioxolane-4-propanol, 2,2-dimethyl-, s,s-3-2,2-dimethyl-1,3-dioxolan-4-yl propan-1-ol,3-4s-2,2-dimethyl-1,3-dioxolan-4-yl-propanol,1,3-dioxolane-4-propanol,2,2-dimethyl-, 4s,3-2,2-dimethyl-1,3-dioxolane-4 s-yl-propanol,3-2,2-dimethyl-1,3-dioxolane-4beta-yl-1-propanol
2733890
3-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]propan-1-ol
CC1(OCC(O1)CCCO)C
51268-87-2
C8H16O3
160.21
MFCD01631673
BSLDYXOPGSOQGZ-ZETCQYMHSA-N
3-4s-2,2-dimethyl-1,3-dioxolan-4-yl propan-1-ol,1,3-dioxolane-4-propanol, 2,2-dimethyl-, s,s-3-2,2-dimethyl-1,3-dioxolan-4-yl propan-1-ol,3-4s-2,2-dimethyl-1,3-dioxolan-4-yl-propanol,1,3-dioxolane-4-propanol,2,2-dimethyl-, 4s,3-2,2-dimethyl-1,3-dioxolane-4 s-yl-propanol,3-2,2-dimethyl-1,3-dioxolane-4beta-yl-1-propanol
2733890
3-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]propan-1-ol
CC1(OCC(O1)CCCO)C
Refractive Index 1.4405 to 1.4425
Boiling Point 90°C (10.0mmHg)
Packaging Glass bottle
Quantity 1g
Specific Rotation +9° to +12° (20°C, 589nm) (c=1, CHCl3)
Color Colorless to Yellow
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 160.21
Physical Form Oily Liquid
Percent Purity 97%
Chemical Name or Material 3-[(4S)-2, 2-Dimethyl-1, 3-dioxolan-4-yl]-propanol, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning